Skip to Content
Merck
All Photos(1)

Key Documents

Z377317

Sigma-Aldrich

Nalgene® Cryoware marker set

black

Synonym(s):

cryomarker, cryopen, freezer marker

Sign Into View Organizational & Contract Pricing

Select a Size

1 PKG
$84.80

$84.80


Available to ship on16 April 2025Details



Select a Size

Change View
1 PKG
$84.80

About This Item

UNSPSC Code:
41103032
NACRES:
NB.35

$84.80


Available to ship on16 April 2025Details


packaging

pkg of 4

manufacturer/tradename

Nalgene 6313-0020

technique(s)

cryopreservation: suitable

color

black

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

For permanently marking cloth, cardboard and plastic. Smudge-proof ink will not fade at ultra-low temperatures. Extra fine tip.

Legal Information

Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

1–2 of 2 Questions  
  1. are these suitable for heat and freezing conditions?

    1 answer
    1. This product is distributed on behalf of ThermoFisher Scientific. It is suitable for cryopreservation. There is no data provided regarding heat resistance.

      Helpful?

  2. are these markers solvent (ethanol) resistant?

    1 answer
    1. This product is not solvent/ethanol resistant. Please see the link below to review product Z744086, which is solvent-resistant and suitable for cryopreservation:
      https://www.sigmaaldrich.com/product/sigma/z744086

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service