HS15994M
Premium slide box
100 place, foam, blue plastic
Synonym(s):
Schiebekasten, slide box microscope, slide storage, slide storage box
Select a Size
$43.00
Available to ship on22 April 2025Details
Select a Size
About This Item
$43.00
Available to ship on22 April 2025Details
Recommended Products
material
blue plastic
nickel closure
feature
Holds 100 x slides 76x26 mm
manufacturer/tradename
Heathrow Scientific HS15994M
L × W × H
208 × 175 × 34 mm
Related Categories
Features and Benefits
- Molded, rigid boxes securely hold slides in grooved slots
- Durable ABS plastic boxes are available with cork lining only
- Both the lining and the slots are numbered to correspond to the slide inventory sheet
- Feature thumb-latch lock mechanism
Choose from one of the most recent versions:
Certificates of Analysis (COA)