Key Documents
HS15994E
Premium slide box
100 place, cork lining, white plastic
Synonym(s):
Schiebekasten, slide box microscope, slide storage, slide storage box
Select a Size
1 EACH
$38.80
$38.80
Available to ship on22 April 2025Details
Select a Size
Change View
1 EACH
$38.80
About This Item
UNSPSC Code:
41123302
NACRES:
NB.17
$38.80
Available to ship on22 April 2025Details
material
nickel closure
white plastic
feature
Holds 100 x slides 76x26 mm
manufacturer/tradename
Heathrow Scientific HS15994E
L × W × H
208 × 175 × 34 mm
Features and Benefits
Slide boxes are perfect for storage or transportation of standard size slides (100 (50 back to back) x 75 mm and 1 x 3").
  • Molded, rigid boxes securely hold slides in grooved slots
  • Durable ABS plastic boxes are available with cork lining only
  • Both the lining and the slots are numbered to correspond to the slide inventory sheet
  • Feature thumb-latch lock mechanism
Choose from one of the most recent versions:
Certificates of Analysis (COA)