Key Documents
F8667
Sigma-Aldrich
Nunc® TripleFlasks, Nunclon® Δ Surface
Culture flask, surface area 500 cm2, with cap filtered, straight neck, sterile, case of 32 ea
Synonym(s):
cell culture flask, tissue culture flask
Select a Size
4 X 8 EA
$1,160.00
$1,160.00
Available to ship on09 April 2025Details
Select a Size
Change View
4 X 8 EA
$1,160.00
About This Item
UNSPSC Code:
41122104
NACRES:
NB.13
$1,160.00
Available to ship on09 April 2025Details
material
screw top blue high-density polyethylene cap
straight neck
straight neck transparent polystyrene flask
sterility
sterile
sterile
packaging
case of 32 ea
manufacturer/tradename
Nunc 132913
flask working volume
200 mL
surface area
500 cm2
500 cm2 , culture area (cm2) per flask
working volume
200 mL
binding type
Nunclon Delta surface
Nunclon Delta surface
General description
Perform cell culture expansion and experience 500cm2 total growth area within the footprint of a standard flask using the Nunc® TripleFlask™ Treated Cell Culture Flasks. Total culture area is achieved by way of the three parallel layers and a straight neck design that works effectively within automation systems. TripleFlask receives the Nunclon™ Delta treatment, a proprietary cell culture surface treatment that offers maximum adhesion for a broad range of cell types. Three layers of growth surfaces provide a total culture area of 500 cm2 in the same footprint as a standard Nunc 175cm2 flask.
Features and Benefits
  • Three parallel growth surfaces provide a total culture area of 500 cm2
  • External dimensions of a 175 cm2 standard flask
  • Ideal for scale-up cell culture applications
  • Extra caps in every carton
  • Nunclon™ Δ surface ensures consistent optimal cell attachment and growth
  • Barcoded with large Code 128
Legal Information
Nunc is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Nunclon is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Choose from one of the most recent versions:
Certificates of Analysis (COA)