Skip to Content
Merck
All Photos(1)

Key Documents

D0446

Sigma-Aldrich

DS1

≥98% (HPLC)

Synonym(s):

4-Chloro-N-[6,8-dibromo-2-(2-thienyl)imidazo[1,2-a]pyridine-3-yl]benzamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H10Br2ClN3OS
CAS Number:
Molecular Weight:
511.62
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

off-white to tan

solubility

DMSO: >10 mg/mL

storage temp.

2-8°C

SMILES string

Clc1ccc(cc1)C(=O)Nc2c(nc3c(Br)cc(Br)cn23)-c4cccs4

InChI

1S/C18H10Br2ClN3OS/c19-11-8-13(20)16-22-15(14-2-1-7-26-14)17(24(16)9-11)23-18(25)10-3-5-12(21)6-4-10/h1-9H,(H,23,25)

InChI key

MEWSBNIVOLYKGU-UHFFFAOYSA-N

Biochem/physiol Actions

δ-GABAA receptors are emerging as an important pharmacological target. DS1 potently (low nM) enhances GABA-evoked currents mediated by α4β3δ receptors. It had little effect on GABA responses mediated by α4β3γ2 receptors. At similar concentrations DS1 directly activates this receptor and is the most potent known agonist of α4β3δ receptors. This compound has an opportunity to be become an agonist golden stardard for δ-GABAA receptors.
DS1 potently (low nM) enhances GABA-evoked currents mediated by α4β3δ receptors.
DS1 potently (low nM) enhances GABA-evoked currents mediated by GABAA α4β3δ receptor subtype. At similar concentrations, it acts as a direct agonist at this receptor and is the most potent agonist known. It has little effect on GABA responses mediated by α4β3γ2 receptors.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiaobo Zhang et al.
PloS one, 7(5), e36822-e36822 (2012-05-17)
The conjunctiva contains a specialized population of lymphocytes that reside in the epithelium, named intraepithelial lymphocytes (IEL). Here we characterized the IEL population prior to and after experimental desiccating stress (DS) for 5 or 10 days (DS5, DS10) and evaluated
Pavel Mazin et al.
Molecular systems biology, 9, 633-633 (2013-01-24)
While splicing differences between tissues, sexes and species are well documented, little is known about the extent and the nature of splicing changes that take place during human or mammalian development and aging. Here, using high-throughput transcriptome sequencing, we have
X Liu et al.
Rice (New York, N.Y.), 11(1), 46-46 (2018-08-08)
Plant height and leaf angle are important determinants of yield in rice (Oryza sativa L.). Genes involved in regulating plant height and leaf angle were identified in previous studies; however, there are many remaining unknown factors that affect rice architecture.
Anne Listrat et al.
Foods (Basel, Switzerland), 8(3) (2019-03-16)
The sensory properties of beef are known to depend on muscle fiber and intramuscular connective tissue composition (IMCT). IMCT is composed of collagens, proteoglycans and glycoproteins. The differentiation of muscle fibers has been extensively studied but there is scarcity in
Nicholas D Huntington et al.
The Journal of experimental medicine, 206(1), 25-34 (2008-12-24)
The in vivo requirements for human natural killer (NK) cell development and differentiation into cytotoxic effectors expressing inhibitory receptors for self-major histocompatibility complex class I (MHC-I; killer Ig-like receptors [KIRs]) remain undefined. Here, we dissect the role of interleukin (IL)-15

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service