Skip to Content
Merck
All Photos(1)

Key Documents

CRM45323

Supelco

Amitraz

certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Synonym(s):

N′-(2,4-Dimethylphenyl)-N-{[(2,4-dimethylphenyl)imino]methyl}-N-methylmethanimidamide, NSC 324552

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
$86.20

$86.20


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 MG
$86.20

About This Item

Empirical Formula (Hill Notation):
C19H23N3
CAS Number:
Molecular Weight:
293.41
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

$86.20


Check Cart for Availability

Request a Bulk Order

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

format

neat

storage temp.

−20°C

SMILES string

CN(\C=N\c1ccc(C)cc1C)\C=N\c2ccc(C)cc2C

InChI

1S/C19H23N3/c1-14-6-8-18(16(3)10-14)20-12-22(5)13-21-19-9-7-15(2)11-17(19)4/h6-13H,1-5H3/b20-12+,21-13+

InChI key

QXAITBQSYVNQDR-ZIOPAAQOSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.Download your certificate at: http://www.sigma-aldrich.com

Application

Amitraz is an amidine acaricide and insecticide for use in top fruit, ornamentals and some vegetables. It also has veterinary treatment applications. It is volatile, almost insoluble in water and, based on its chemical properties, is only slightly mobile with a low potential for leaching to groundwater. As per the regulation (EC) No 1107/2009 it is not approved for use as plant protection product in the European Union. Maximum residuel levels (MRLs) have been set for amitraz (including the metabolites containing the 2,4-dimethylaniline moiety expressed as amitraz) according to Reg. (EU) 2017/623 for various products of plant and animal origin from 0.01 - 0.4 mg/kg . The certified reference material (CRM) is intended to be used as a calibrant for chromatography and other analytical techniques.
Environmental

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Skin Sens. 1 - STOT RE 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service