Skip to Content
Merck
All Photos(1)

Key Documents

CLS3886

Corning® 96 Well Half-Area Microplate

flat bottom clear, white polystyrene, Tissue Culture (TC)-treated surface, bag of 20, sterile, lid: no

Synonym(s):

96 multiwell plate

Sign Into View Organizational & Contract Pricing

Select a Size

100 EA
$2,090.00

$2,090.00


Check Cart for Availability


Select a Size

Change View
100 EA
$2,090.00

About This Item

UNSPSC Code:
41121800
NACRES:
NB.15
material:
flat bottom clear
round wells
white polystyrene
size:
96 wells

$2,090.00


Check Cart for Availability

material

flat bottom clear
round wells
white polystyrene

sterility

sterile

feature

lid: no
skirt

packaging

bag of 20
pkg of 20 inner package

manufacturer/tradename

Corning 3886

size

96 wells

well D

10.7 mm

well volume

190 μL

well working volume

25-125 μL

color

white

suitability

suitable for (cell culture)

binding type

Tissue Culture (TC)-treated surface

Looking for similar products? Visit Product Comparison Guide

General description

Unique half area plates can help save on rare and expensive reagents by reducing the amount of reagent needed per well, while still being readable by most standard plate readers.

Legal Information

Corning is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service