Skip to Content
Merck
All Photos(1)

Documents

GE28-9068-36

Hyperfilm ECL

W × L 18 cm × 24 cm, pack of 50 ea

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105300
NACRES:
NB.22

form

solid

feature

wetted part: no

packaging

pack of 50 ea

manufacturer/tradename

Cytiva 28906836

W × L

18 cm × 24 cm

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Hyperfilm ECL film is double coated. It provides improved clarity that gives excellent contrast and band visibility in the detection of chemiluminescent signals on Western blot analysis.

Hyperfilm ECL film provides low background with reduction in visible artifacts, and excellent low concentration target detection. Hyperfilm ECL film is suitable for use with all blue and green light chemiluminescent systems and validated for use with ECL start reagent, ECL reagent, ECL Prime reagent, ECL Select reagent, and CDP-Star reagent.

Hyperfilm ECL film is constructed from a clear base coated on both sides with a photographic emulsion that is protected by an anti-scratch layer. The anti-scratch coating reduces the risk of visible artifacts (e.g., plastic wrap marks), and the clear colorless base results in easy reading of images without compromising on sensitivity.

Improve detection of lower concentration targets during chemiluminescence Western blot analysis by extending exposure times, without increasing background noise. Hyperfilm ECL film can be processed in automatic processors or manually in tanks and trays using most common x-ray film developers and fixers.

Application

Hyperfilm ECL has been used:
  • to produce an image fit for publication
  • for blue and green light-based chemiluminescence northern, southern, and western blotting procedures
  • to identify proteins and nucleic acids at low concentrations
  • in immunoprecipitation

Features and Benefits

  • High-sensitivity Western blot membrane to detect low concentrations of nucleic acid and protein targets
  • Optimized for use in all blue and green light chemiluminescence Western blot systems
  • Clear background for excellent contrast and improved band resolution
  • Produces publication-quality images
  • Easy to read
  • Simple to use

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.

Legal Information

ECL is a trademark of Cytiva
Hyperfilm is a trademark of Cytiva

Storage Class Code

11 - Combustible Solids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lucie M Wolf et al.
Journal of cell science, 134(16) (2021-08-19)
WNT signalling is important for development in all metazoans and is associated with various human diseases. The ubiquitin-proteasome system (UPS) and regulatory endoplasmic reticulum-associated degradation (ERAD) have been implicated in the production of WNT proteins. Here, we investigated how the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service