CLS3770BC
Corning® CellBIND® 384 well microplate
CellBIND® surface, black polystyrene, flat bottom, sterile, with generic barcode, lid, pack of 20
Synonym(s):
384 multiwell plates
Select a Size
$878.00
Select a Size
About This Item
black polystyrene
clear bottom
flat bottom
$878.00
Recommended Products
material
black plate
black polystyrene
clear bottom
flat bottom
sterility
sterile
feature
lid
skirt
Low Flange
with generic barcode
packaging
case of 100 plates
pack of 20
manufacturer/tradename
Corning 3770BC
size
384 wells
well volume
112 μL
well working volume
25-50 μL
wells
384 wells
suitability
suitable for (cell cuture)
binding type
CellBIND® surface
Related Categories
Features and Benefits
- A flat, optically clear well bottom enables direct microscopic examination
- Low background fluorescence and opaque walls minimize crosstalk between wells and light scattering
- Wells are identified using individual alphanumeric codes
- Gamma radiation sterilized and nonpyrogenic
- Corning® CellBIND® treated surface for better cell attachment
- Suitable for the bottom and top reading instruments
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)