CLS3264
Corning® CellCube® Module 100 Layer with 85,000 cm2 growth surface
TC-Treated, square flask, sterile
Synonym(s):
cell culture vessel
Select a Size
$6,050.00
Select a Size
About This Item
$6,050.00
Recommended Products
material
square bottom flask
sterility
sterile
feature
cap: no
square flask
packaging
case of 1 ea
manufacturer/tradename
Corning
flask surface area
(85,000 cm2)
binding type
Tissue Culture (TC)-treated surface
Related Categories
General description
These CellCube Modules provide:
- A series of parallel, polystyrene plates joined to create thin, sealed, laminar flow spaces between adjacent plates
- 85,000 cm2 polystyrene growth surface optimally treated for the growth of attachment dependent cells
- A perfused environment which more closely simulates in vivo conditions and reliably distributes nutrients and oxygen
- An integral, encapsulated, sterile, single-use device that is 100% pressure-hold tested before shipment
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service