CLS3233
Corning® CellCube®
100 layer, TCT, with 6” Silicone tubing and Aseptic Connectors, sterile, bag of 1 ea, case of 1 ea
Synonym(s):
cell culture vessel
Select a Size
$6,760.00
Select a Size
About This Item
$6,760.00
Recommended Products
material
polystyrene
silicon tubing (6′′)
sterility
sterile
packaging
bag of 1 ea
case of 1 ea
manufacturer/tradename
Corning 3233
surface area
8500 cm2 , cell growth area
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Application
Features and Benefits
- 8,500 cm2 Surface Area
- Corning TCT surface treatment
- Sterility Assurance Level(SAL) of 10-6
- AseptiQuik® G Connector
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service