CLS3270
Corning® CellSTACK® cell culture chambers
Tissue Culture (TC)-treated surface, clear polystyrene, sterile, graduations, high-density polyethylene cap, case of 2
Synonym(s):
cell culture flasks, scale-up cell culture flasks, tissue culture flasks
About This Item
material
clear polystyrene
high-density polyethylene cap
rectangular bottom flask
short neck
sterility
sterile
feature
cap vented
graduations
packaging
pack of 1
case of 2
manufacturer/tradename
Corning 3270
surface area
6360 cm2
working volume
1250-1900 mL
suitability
suitable for (scale-up cell culture)
binding type
Tissue Culture (TC)-treated surface
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
- Ideal for cell culture scale-up
- Culture chambers produce cell yields in the 107 - 109 range
- Useful alternatives to multiple roller bottles or spinner flasks.
- High mechanical strength and structural integrity
- 100% leak tested prior to shipping
- Certified nonpyrogenic and sterilized by gamma irradiation, with low particulate contamination from modern manufacturing methods
- Easy to use larger openings with threaded closures and vented caps
- Large 26 mm wide filling ports allow direct access to chamber bottom, with more options for sterile transfer of media and cultures
Features and Benefits
- Has 6,360cm2 cell growth area
- Two 26mm diameter filling ports allow direct access to chamber bottom providing greater flexibility for sterile filling and emptying by pouring, pipetting or via tubing in a fully closed system
- Standard 33mm threaded caps have 0.2μm pore nonwettable membranes sealed directly to the caps to allow gas exchange while minimizing the risk of contamination
- Optional 33mm threaded caps are available with integrally sealed USP Class VI certified C-FLEX tubing to allow direct sterile transfer of media and cells
- USP Class VI polystyrene provides excellent optical clarity and mechanical strength
- Sterilized by gamma radiation and certifiied nonpyrogenic
- 100% leak tested prior to shipping
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service