Skip to Content
Merck
All Photos(1)

Key Documents

CLS3099

Corning® multiwell plates, plate lids and sealing mats

universal multiwell plate lid w/ corner notch, polystyrene, clear, sterile, 50/cs

Synonym(s):

universal microplate lid, universal microtiter plate lid, universal multiwell plate lid, universal plate lid

Sign Into View Organizational & Contract Pricing

Select a Size

50 EA
$105.00

$105.00


Estimated to ship on22 March 2025



Select a Size

Change View
50 EA
$105.00

About This Item

UNSPSC Code:
41121800
NACRES:
NB.13
material:
polystyrene
size:
96 or 384 wells

$105.00


Estimated to ship on22 March 2025


material

polystyrene

sterility

sterile

feature

lid: no

packaging

pack of 25
case of 50

manufacturer/tradename

Corning 3099

size

96 or 384 wells

color

clear

Looking for similar products? Visit Product Comparison Guide

General description

  • Lids are made of rigid polystyrene
  • Lids have a corner notch on the A1 corner to correspond to the corner notches found on all Corning® microplates
  • Suitable for 96 well, 96 half area & 384 well plates
  • Lids do not fit Corning 2 μL 1536 well microplates
  • Sterilized by gamma radiation

Legal Information

Corning is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service