Skip to Content
Merck
All Photos(1)

Documents

CLS3792

Corning® 96 Well Solid Polystyrene Microplate

round bottom, black polystyrene, bag of 25, non-sterile, lid: no

Synonym(s):

96 microplates, 96 microtiter plates, 96 multiwell plates, 96 well plates

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.15

material

black polystyrene
round bottom

sterility

non-sterile

feature

lid: no

packaging

bag of 25

manufacturer/tradename

Corning 3792

size

96 wells

well volume

330 μL

working volume

75-200 μL

color

black

binding type

non-treated surface

Looking for similar products? Visit Product Comparison Guide

General description

  • Corning® 96 Well Solid Polystyrene Microplate round bottom is black polystyrene, not treated ELISA microplate.
  • Displays low background fluorescence and minimizes light scattering
  • Minimal well-to-well crosstalk
  • Hydrophobic surface and binds biomolecules through passive interactions
  • Applicable for the immobilization of large molecules
  • Binding capacity of approximately 100 to 200ng IgG/cm²
  • Standard microplate footprint and dimensions

Legal Information

Corning is a registered trademark of Corning, Inc.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service