Skip to Content
Merck
All Photos(1)

Key Documents

CLS139780LTC

Corning® GLS 80 polypropylene screw cap with plug seal

orange cap, non-sterile, case of 20

Synonym(s):

Corning screw cap

Sign Into View Organizational & Contract Pricing

Select a Size

20 EA
$129.00

$129.00


Check Cart for Availability


Select a Size

Change View
20 EA
$129.00

About This Item

UNSPSC Code:
41121800

$129.00


Check Cart for Availability

sterility

non-sterile

packaging

case of 20

manufacturer/tradename

Corning 1397-80LTC

color

orange cap

General description

  • Easy on-off quick relaease cap
  • Cap is sealed by using an integral plug designed to mold against the inside rim of the bottle
  • Audible click once the cap is sealed on the bottle

Legal Information

Corning is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service