Skip to Content
Merck
All Photos(1)

Key Documents

CLS139545SS

Corning® Silicone Septa for GL45 Screw Cap

Synonym(s):

Corning cap

Sign Into View Organizational & Contract Pricing

Select a Size

10 EA
$88.00

$88.00


Check Cart for Availability


Select a Size

Change View
10 EA
$88.00

About This Item

UNSPSC Code:
41121800
NACRES:
NB.43

$88.00


Check Cart for Availability

sterility

non-sterile

feature

cap: no

manufacturer/tradename

Corning 139545SS

color

natural

General description

The Corning silicone septa fit into open top PBT caps (No. CLS139545HTSC) with GL45 threads to allow for aseptic reagent delivery or culture sampling using a syringe and needle. They can be autoclaved at 121°C or dry heat sterilized at up to 180°C.

Legal Information

Corning is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service