Skip to Content
Merck
All Photos(2)

Key Documents

CLS139525SS

Corning® Silicone Septa

for use with GL25 Open Top PBT Screw Cap

Synonym(s):

Corning cap

Sign Into View Organizational & Contract Pricing

Select a Size

10 EA
$26.70

$26.70


Available to ship on29 April 2025Details



Select a Size

Change View
10 EA
$26.70

About This Item

UNSPSC Code:
41103800

$26.70


Available to ship on29 April 2025Details


material

natural

sterility

non-sterile

packaging

pack of 10

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

The Corning silicone septa fit into open top PBT caps (CLS139525HTSC) with GL25 threads to allow for aseptic reagent delivery or culture sampling using a syringe and needle. They can be autoclaved at 121deg. C or dry heat sterilized at up to 180 deg. C.

Legal Information

Corning is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service