Skip to Content
Merck
All Photos(1)

Key Documents

CLS139545HTSC

Corning® high-temp PBT screw cap

For high temps, red, GL 45 open-top

Synonym(s):

Corning screw cap

Sign Into View Organizational & Contract Pricing

Select a Size

10 EA
$97.70

$97.70


Available to ship on22 April 2025Details



Select a Size

Change View
10 EA
$97.70

About This Item

UNSPSC Code:
41121800
NACRES:
NB.43

$97.70


Available to ship on22 April 2025Details


material

PBT

manufacturer/tradename

Corning 1395-45HTSC

color

red

fitting

GL 45 open-top

Looking for similar products? Visit Product Comparison Guide

Legal Information

Corning is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service