Skip to Content
Merck
All Photos(1)

Key Documents

C6267

Supelco

Chromosorb® LC-7

10 μm

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
$986.00

$986.00


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
10 G
$986.00

About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12000000
NACRES:
SB.52

$986.00


Estimated to ship on28 May 2025


Request a Bulk Order

form

solid

surface area

>250 m2/g

particle size

10 μm

density

0.45 g/cm3 (lit.)

Looking for similar products? Visit Product Comparison Guide

Application

A reverse phase microparticulate packing used for separating a variety of compounds used in pharmaceutical and food products.

Legal Information

Chromosorb is a registered trademark of Imerys Minerals California, Inc.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Carc. 2 - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Weronika Kuźmicka et al.
Nutrients, 13(1) (2020-12-31)
Zinc plays an important physiological role in the entire body, especially in the immune system. It is one of the most abundant microelements in our organism and an essential component of enzymes and antibacterial proteins. Zinc levels were reported to

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service