Skip to Content
Merck
All Photos(1)

Key Documents

C3527

Sigma-Aldrich

Anti-C-Reactive Protein antibody produced in rabbit

IgG fraction of antiserum, lyophilized powder

Synonym(s):

Anti-CRP

Sign Into View Organizational & Contract Pricing

Select a Size

1 VIAL
$537.00

$537.00


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
1 VIAL
$537.00

About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46

$537.00


Estimated to ship on28 May 2025


Request a Bulk Order

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

lyophilized powder

species reactivity

human

packaging

vial of 2 mL lyophilized antiserum

technique(s)

indirect ELISA: 1:10,000-1:20,000

UniProt accession no.

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... CRP(1401)

Related Categories

General description

C-reactive protein (CRP) is an acute-phase pentraxin protein. It is synthesized by the liver. This gene is mapped to human chromosome 1q23. It has two exons and an intron. It is located within the complement membrane complex in early atherosclerotic tissue.

Application

Anti-C-Reactive Protein antibody produced in rabbit has been used in electro immunoassay, and enzyme-linked immunosorbent assay (ELISA).
Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Enzyme-linked immunosorbent assay (1 paper)

Biochem/physiol Actions

C-reactive protein (CRP) is considered as a major biomarker of sterile inflammation. It is mainly used to observe the treatment response which can predict long-term consequence in inflammatory diseases. CRP facilitates opsonization of low-density lipoprotein by macrophages.

Physical form

Lyophilized from 0.01 M phosphate buffered saline, pH 7.2

Reconstitution

Reconstitute with 2 ml deionized water.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C-Reactive Protein Production in Term Human Placental Tissue
Malek A, et al.
Placenta, 27(6), 619-625 (2006)
Protein and amino acid metabolism in three-to twelve-month-old infants fed human milk or formulas with varying protein concentrations
AAkeson PMK, et al.
Journal of Pediatric Gastroenterology and Nutrition, 26(3), 297-304 (1998)
Luigi Raio et al.
Journal of hypertension, 37(5), 1012-1017 (2019-03-19)
Angiogenic and inflammatory factors have been shown to play an important role in the pathogenesis of preeclampsia. However, there is little information on their interaction. The aims of this study were to investigate the longitudinal pattern of inflammatory markers, such
Relation of polymorphism within the C-reactive protein gene and plasma CRP levels
Suk HJ, et al.
Atherosclerosis, 178(1), 139-145 (2005)
C-Reactive Protein Levels at the Midpregnancy Can Predict Gestational Complications
Vecchie A, et al.
BioMed Research International, 2018 (2018)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service