Skip to Content
Merck
All Photos(1)

Key Documents

OR03L

Sigma-Aldrich

Streptavidin, Horseradish Peroxidase Conjugate

This Streptavidin, Horseradish Peroxidase Conjugate is validated for use in ELISA, Immunoblotting, Immunohistochemistry for the detection of Streptavidin-Horseradish Peroxidase Conjugate.

Synonym(s):

Biotin-Binding Protein

Sign Into View Organizational & Contract Pricing

Select a Size

200 μG
$155.00

$155.00


Estimated to ship on13 April 2025


Request a Bulk Order

Select a Size

Change View
200 μG
$155.00

About This Item

UNSPSC Code:
41116133
NACRES:
NA.41

$155.00


Estimated to ship on13 April 2025


Request a Bulk Order

Quality Level

form

lyophilized

does not contain

preservative

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

shipped in

ambient

storage temp.

−20°C

General description

Streptavidin conjugated to horseradish peroxidase.

Application

ELISA (10-200 ng/ml)

Immunoblotting (10-200 ng/ml)

Immunohistochemistry (10-200 ng/ml)

Warning

Toxicity: Standard Handling (A)

Physical form

Lyophilized from 250 mM NaCl, 10 mM sodium phosphate, 15 mg/ml BSA, pH 7.6.

Reconstitution

Reconstitute the lyophilized conjugate with in 1 ml sterile dH₂O. Lyophilized conjugate should be resuspended at 4°C with occasional gentle mixing for at least 2 h. Following reconstitution, aliquot and freeze (-20°C). Avoid freeze/thaw cycles of solutions.

Other Notes

Direct conjugates of streptavidin are provided to be used in combination with biotinylated Calbiochem monoclonal antibodies. Conjugates should be titrated for optimal results in individual systems.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service