Skip to Content
Merck
All Photos(1)

Key Documents

AXYRFLR1200

Empty Pipette tip refill rack, with lid

for use with 1200 μL tip inserts, pkg of 3x10racks/cs

Synonym(s):

pipet tips

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122800
NACRES:
NB.02
packaging:
pkg of 3x10racks/cs
manufacturer/tradename:
Axygen Inc RFL-R-1200
Corning
Product AXYRFLR1200 is not currently sold in your country. Contact Technical Service

manufacturer/tradename

Axygen Inc RFL-R-1200
Corning

packaging

pkg of 3x10racks/cs

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service