Skip to Content
Merck
All Photos(1)

Key Documents

A6781

Sigma-Aldrich

Acetate Kinase from Bacillus stearothermophilus

lyophilized powder, 400-1,200 units/mg solid

Synonym(s):

ATP:Acetate phosphotransferase, Acetate Kinase Bacillus stearothermophilus

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
MDL number:
UNSPSC Code:
12352204
eCl@ss:
32160410

biological source

bacterial (Bacillus stearothermophilus)

Quality Level

form

lyophilized powder

specific activity

400-1,200 units/mg solid

storage temp.

2-8°C

Application

Acetate kinase is used to phosphorylate acetate to acetyl phosphate. Acetate Kinase from Bacillus stearothermophilus has been used to study allosteric activation.[1] [32P]-acetyl phosphate was generated by incubating potassium acetate in the reaction mixture with acetate kinase from Sigma. This [32P]-acetyl phosphate was used to label BldM, BldM D-54N or BldM D-54A loci during the study of the effect of bldM gene on Streptomyces coelicolor development.[2]

Biochem/physiol Actions

Acetate kinase plays an important role in glycolysis. Acetate kinase phosphorylates acetate in the presence of ATP and a divalent cation, which ultimately results in the production of acetyl-CoA.[3] Acetate kinase is also involved in the metabolism of propanoate, pyruvate and taurine. Acetate Kinase from Bacillus stearothermophilus is a thermostable tetramer of identical subunits with molecular weight of 40,000 Da each. The enzyme does not have a -SH group and is composed of 36% β-structure, 21 % α-helix and 43 % unordered structure.[4]
Involved in the metabolism of propanoate, pyruvate and taurine.

Unit Definition

One unit will phosphorylate 1.0 μmole of acetate to acetyl phosphate per min at pH 7.2 at 30 °C.

Physical form

Contains Tris-HCl buffer.

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H Nakajima et al.
Journal of biochemistry, 86(5), 1169-1177 (1979-11-01)
Fructose 1,6-bisphosphate (FBP) stimulates the reaction of Bacillus stearothermophilus acetate kinase (AK). FBP changes the reaction curve for ATP from a sigmoidal type to a Michaelis-Menten one. The binding of FBP to AK was studied by an equilibrium dialysis method
H Nakajima et al.
Journal of biochemistry, 84(1), 193-203 (1978-07-01)
1. Acetate kinase [EC 2.7.2.1] from an thermophile, B. stearothermophilus, was purified and crystalized. 2. This enzyme was shown to be a tetramer of identical subunits which had a molecular weight of about 40,000. Amino acid analysis showed no SH
Sagar Chittori et al.
Acta crystallographica. Section F, Structural biology and crystallization communications, 67(Pt 12), 1658-1661 (2011-12-06)
Acetate kinase (AckA) catalyzes the reversible transfer of a phosphate group from acetyl phosphate to ADP, generating acetate and ATP, and plays a central role in carbon metabolism. In the present work, the gene corresponding to AckA from Salmonella typhimurium
Kuo-Hsiang Tang et al.
The Journal of biological chemistry, 285(46), 35848-35854 (2010-07-24)
The anoxygenic green sulfur bacteria (GSBs) assimilate CO(2) autotrophically through the reductive (reverse) tricarboxylic acid (RTCA) cycle. Some organic carbon sources, such as acetate and pyruvate, can be assimilated during the phototrophic growth of the GSBs, in the presence of
K A Buss et al.
Journal of bacteriology, 183(2), 680-686 (2001-01-03)
Acetate kinase, an enzyme widely distributed in the Bacteria and Archaea domains, catalyzes the phosphorylation of acetate. We have determined the three-dimensional structure of Methanosarcina thermophila acetate kinase bound to ADP through crystallography. As we previously predicted, acetate kinase contains

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service