Skip to Content
Merck
All Photos(1)

Key Documents

HUMANCD14-0002081

Sigma-Aldrich

Frozen Human Normal CD14+ Monocytes

human peripheral blood (monocytes)

Synonym(s):

CD14+ Cells, Immune Cells, Monocytes

Sign Into View Organizational & Contract Pricing

Select a Size

1 VIAL
$1,620.00

$1,620.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 VIAL
$1,620.00

About This Item

UNSPSC Code:
41106514

$1,620.00


Check Cart for Availability

Request a Bulk Order

Product Name

Frozen Human Normal CD14+ Monocytes, 10 Million Cells/per vial

biological source

human peripheral blood (monocytes)

Quality Level

form

frozen liquid

packaging

vial of 10 Million cells

manufacturer/tradename

BioIVT

technique(s)

cell culture | mammalian: suitable

color

cloudy

storage temp.

-140 to -196°C

General description

Lot specific orders are not able to be placed through the web. Contact your local sales rep for more details.

We are pleased to offer these products from BioIVT.

Monocytes are a subset of leukocytes, or white blood cells, found in peripheral blood and can migrate into tissue to differentiate into macrophages or myeloid lineage dendritic cells. CD14+ Monocytes are isolated from peripheral blood and separated from mononuclear cells via immunomagnetic separation.

Used for a variety of applications, monocytes are frequently utilized in drug discovery/development, assay validation/development, monocyte activation tests, and other immunology applications.

Packaging

10 Million Cells/per vial

Other Notes

Thaw before use.

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Met. Corr. 1 - Skin Irrit. 2

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. Are these monocytes pooled or from individual donors? We're looking for a lot of pooled CD14+ human monocytes for our lab.

    1 answer
    1. They are pooled from individual donor sample.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service