A1233
AMPK (α1/β1/γ1), active, His tagged human
PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution
Synonym(s):
AMPKG, HAMPKb, MGC17785, MGC33776, MGC57364, MGC8666, PRKAA1, PRKAB1, PRKAG1
Select a Size
About This Item
Recommended Products
biological source
human
Quality Level
recombinant
expressed in baculovirus infected Sf9 cells
product line
PRECISIO® Kinase
Assay
≥70% (SDS-PAGE)
form
buffered aqueous glycerol solution
specific activity
626-848 nmol/min·mg
mol wt
A1 subunit ~68 kDa
B1 subunit ~38 kDa
G1 subunit ~40 kDa
solubility
water: soluble
application(s)
life science and biopharma
shipped in
dry ice
storage temp.
−70°C
Gene Information
human ... PRKAA1(5562) , PRKAB1(5564) , PRKAG1(5571)
General description
AMP-activated protein kinase (AMPK) is a heterotrimer protein kinase consisting of a α catalytic subunit, and non-catalytic β and γ subunits.[1]
Application
Biochem/physiol Actions
Physical form
Legal Information
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Repr. 1B - Skin Irrit. 2
Storage Class Code
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service