Skip to Content
Merck
All Photos(1)

Key Documents

171536

Sigma-Aldrich

AMPK (α1/β1/γ1), His•Tag®, Human, Recombinant, S. frugiperda

full length enzyme consisting of subunits A1, B1, and G1

Sign Into View Organizational & Contract Pricing

Select a Size

5 μG
$952.00

$952.00


Estimated to ship on31 May 2025


Request a Bulk Order

Select a Size

Change View
5 μG
$952.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.77

$952.00


Estimated to ship on31 May 2025


Request a Bulk Order

recombinant

expressed in insect cells (S. frugiperda)

Quality Level

Assay

≥95% (SDS-PAGE)

form

liquid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

shipped in

wet ice

storage temp.

−70°C

General description

Full length, recombinant, human AMPK subunits (A1, B1, and G1), each fused at the C-terminus to a His•Tag sequence and co-expressed in S. frugiperda insect cells using a baculovirus expression system. AMPK is involved in sensing energy levels in the cell that result from changes in AMP levels. AMPK is an upstream regulator of mTOR activation. The active complex is useful in the study of kinase activity regulation and inhibitor screening. M.W. = 68,000 (A1), 38,000 (B1), 40,000 (G1).
M.W. = 68,000 (A1), 38,000 (B1), 40,000 (G1)

Warning

Toxicity: Standard Handling (A)

Physical form

In 300 mM NaCl, 150 mM Imidazole, 50 mM Sodium Phosphate, 0.2 mM DTT, 0.1 mM PMSF, 25% glycerol, pH 7.0.

Reconstitution

Following initial thaw, aliquot and freeze (-70°C).

Other Notes

Scott J.W., 2007. EMBO J26, 806.
Minokoshi Y et al. 2004. Nature428, 569.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
HIS TAG is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service