Skip to Content
Merck
All Photos(1)

Key Documents

05438

Sigma-Aldrich

Albumin from chicken egg white

lyophilized powder, ≥90% (agarose gel electrophoresis)

Synonym(s):

Albumin from hen egg white, Ovalbumin

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
$264.00

$264.00


Check Cart for Availability


Select a Size

Change View
500 MG
$264.00

About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352202
eCl@ss:
42010201

$264.00


Check Cart for Availability

biological source

chicken egg white

Assay

≥90% (agarose gel electrophoresis)

form

lyophilized powder

mol wt

44287 Da by calculation

technique(s)

electrophoresis: suitable

UniProt accession no.

storage temp.

−20°C

Gene Information

chicken ... OVAL(396058)

Looking for similar products? Visit Product Comparison Guide

Application

A phosphorylated glycoprotein which is useful as a marker (~45 kDa) for protein electrophoresis. Has been used to produce an animal model of asthma.

Preparation Note

from crystallized, lyophilized and desalted ovalbumin.

Reconstitution

0.9-1.4 mg/mL after reconstitution with 1 mL of water

Other Notes

Sales restrictions may apply

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Resp. Sens. 1A

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marina Pereira De Campos et al.
Biological & pharmaceutical bulletin, 28(8), 1527-1530 (2005-08-05)
We have studied the crude methanolic extract (CME), some fractions (hexane, dichloromethane and ethyl acetate) and four pure compounds: eupomatenoid-3 (1), eupomatenoid-5 (2), conocarpan (3) and orientin (4), from Piper solmsianum, for possible antifungal activity against 12 pathogenic fungi. The
Preety Sahdev et al.
Journal of biomedical nanotechnology, 9(1), 132-141 (2013-05-01)
The main objective of this study was to investigate the potential of calcium phosphate (CAP) nanoparticles for transcutaneous vaccine delivery. CAP nanoparticles were prepared by nanoprecipitation method followed by sequential adsorption of sugars and ovalbumin. Nanoparticles were characterized using dynamic
Simon Heidegger et al.
Journal of immunology (Baltimore, Md. : 1950), 190(10), 5313-5320 (2013-04-17)
The trafficking of effector T cells is tightly regulated by the expression of site-specific sets of homing molecules. In contrast, naive T cells are generally assumed to express a uniform pattern of homing molecules and to follow a random distribution
Eniko Lele et al.
Anesthesia and analgesia, 116(6), 1257-1264 (2013-04-18)
Volatile inhaled anesthetics exert a differential protective effect against bronchospasm development after cholinergic stimulation. However, their ability to inhibit the adverse respiratory consequences of an anaphylactic reaction after exposure to an allergen has not been characterized. We therefore compared the
Qing-Rong Tuo et al.
Biochemical and biophysical research communications, 434(3), 498-502 (2013-04-16)
The level of reactive oxygen species (ROS) and the activity of spontaneous, transient, localized Ca(2+) increases (known as Ca(2+) sparks) in tracheal smooth muscle cells (TSMCs) in an experimental allergic asthma mouse model has not yet been investigated. We used

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service