Merck
  • TLR activation excludes circulating naive CD8+ T cells from gut-associated lymphoid organs in mice.
TLR activation excludes circulating naive CD8+ T cells from gut-associated lymphoid organs in mice.
Journal of immunology (Baltimore, Md. : 1950) (2013-04-17)
Simon Heidegger, Sophie-Kathrin Kirchner, Nicolas Stephan, Bernadette Bohn, Nina Suhartha, Christian Hotz, David Anz, Nadja Sandholzer, Bärbel Stecher, Holger Rüssmann, Stefan Endres, Carole Bourquin
ABSTRACT
The trafficking of effector T cells is tightly regulated by the expression of site-specific sets of homing molecules. In contrast, naive T cells are generally assumed to express a uniform pattern of homing molecules and to follow a random distribution within the blood and secondary lymphoid organs. In this study, we demonstrate that systemic infection fundamentally modifies the trafficking of circulating naive CD8(+) T cells. We show that on naive CD8(+) T cells, the constitutive expression of the integrin α4β7 that effects their entry into GALT is downregulated following infection of mice with Salmonella typhimurium. We further show that this downregulation is dependent on TLR signaling, and that the TLR-activated naive CD8(+) T cells are blocked from entering GALT. This contrasts strongly with Ag-experienced effector T cells, for which TLR costimulation in the GALT potently upregulates α4β7 and enhances trafficking to intestinal tissues. Thus, TLR activation leads to opposite effects on migration of naive and effector CD8(+) T cells. Our data identify a mechanism that excludes noncognate CD8(+) T cells from selected immune compartments during TLR-induced systemic inflammation.
Sigma-Aldrich
Albumin from chicken egg white, lyophilized powder, ≥90% (agarose gel electrophoresis)
Sigma-Aldrich
Albumin from chicken egg white, lyophilized powder, ≥98% (agarose gel electrophoresis)
Supelco
Albumin from chicken egg white, For use as a marker in SDS-PAGE
Sigma-Aldrich
Albumin from chicken egg white, lyophilized powder
Sigma-Aldrich
Albumin from chicken egg white, lyophilized powder, ≥98% (agarose gel electrophoresis)
Sigma-Aldrich
Albumin from chicken egg white, lyophilized powder, ≥90% (agarose gel electrophoresis)