Skip to Content
Merck
All Photos(1)

Key Documents

Y0001023

Nicotine for system suitability

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

(−)-Nicotine hydrogen tartrate salt, (−)-1-Methyl-2-(3-pyridyl)pyrrolidine (+)-bitartrate salt

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$254.00

$254.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 MG
$254.00

About This Item

Empirical Formula (Hill Notation):
C10H14N2 · 2C4H6O6
CAS Number:
Molecular Weight:
462.41
UNSPSC Code:
41116107
NACRES:
NA.24

$254.00


Check Cart for Availability

Request a Bulk Order

grade

pharmaceutical primary standard

API family

nicotine

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

−20°C

SMILES string

CN1CCC[C@H]1C2=CC=CN=C2.C

InChI

1S/C10H14N2.CH4/c1-12-7-3-5-10(12)9-4-2-6-11-8-9;/h2,4,6,8,10H,3,5,7H2,1H3;1H4/t10-;/m0./s1

InChI key

RYYKFQGHMYGLEL-PPHPATTJSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Nicotine for system suitability EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

Prototype nicotinic acetylcholine receptor agonist; naturally occurring isomer.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 2 - Eye Irrit. 2 - Repr. 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 2

Flash Point(F)

289.8 °F

Flash Point(C)

143.2 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Matthew L Banks
Experimental and clinical psychopharmacology, 22(3), 266-273 (2014-02-20)
Preclinical drug discrimination procedures have been useful in understanding the pharmacological mechanisms of the subjective-like effects of abused drugs. Converging lines of evidence from neurochemical and behavioral studies implicate a potential role of nicotinic acetylcholine (nACh) receptors in the abuse-related
June Bryan de la Peña et al.
Behavioural brain research, 272, 156-164 (2014-07-06)
Nicotine/cigarette addiction starts young. Indeed, most smokers started when they were adolescents. Adolescence has been implicated to be a critical period for nicotine/cigarette addiction, thus it is important to understand the consequences of such early exposure. In the present study
Yuan-yuan Zhang et al.
Acta pharmacologica Sinica, 35(9), 1137-1149 (2014-08-05)
The non-neuronal acetylcholine system (NNAS) in endothelial cells participates in modulating endothelial function, vascular tone, angiogenesis and inflammation, thus plays a critical role in cardiovascular diseases. In this study, we used a proteomic approach to study potential downstream receptor-effectors of
Raphaël Eddine et al.
Scientific reports, 5, 8184-8184 (2015-02-03)
Midbrain dopamine (DA) neurons are key players in motivation and reward processing. Increased DA release is thought to be central in the initiation of drug addiction. Whereas dopamine neurons are generally considered to be activated by drugs such as nicotine
P P Muldoon et al.
British journal of pharmacology, 172(3), 869-882 (2014-09-27)
Abrupt discontinuation of nicotine, the main psychoactive component in tobacco, induces a withdrawal syndrome in nicotine-dependent animals, consisting of somatic and affective signs, avoidance of which contributes to drug maintenance. While blockade of fatty acid amide hydrolase, the primary catabolic

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service