![Nicotine for system suitability European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/379/324/b1ed15bc-a901-4843-b31f-097377a6494e/640/b1ed15bc-a901-4843-b31f-097377a6494e.png)
Nicotine for system suitability
![Nicotine for system suitability European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/379/324/b1ed15bc-a901-4843-b31f-097377a6494e/640/b1ed15bc-a901-4843-b31f-097377a6494e.png)
Synonym(s):
(−)-Nicotine hydrogen tartrate salt, (−)-1-Methyl-2-(3-pyridyl)pyrrolidine (+)-bitartrate salt
Empirical Formula (Hill Notation):
C10H14N2 · 2C4H6O6
CAS Number:
Molecular Weight:
462.41
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing