Skip to Content
Merck
All Photos(1)

Key Documents

Y0001425

Nicotine ditartrate dihydrate

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

(−)-Nicotine hydrogen tartrate salt dihydrate

Sign Into View Organizational & Contract Pricing

Select a Size

15 MG
$254.00

$254.00


Available to ship on14 April 2025Details


Request a Bulk Order

Select a Size

Change View
15 MG
$254.00

About This Item

Empirical Formula (Hill Notation):
C10H14N2 · 2C4H6O6 · 2H2O
CAS Number:
Molecular Weight:
498.44
UNSPSC Code:
41116107
NACRES:
NA.24

$254.00


Available to ship on14 April 2025Details


Request a Bulk Order

grade

pharmaceutical primary standard

API family

nicotine

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

Clc1cc2c(cc1)O\C(=C3\C=C(C=CC\3=O)NC(=O)C(C)C)\N2

InChI

1S/C17H15ClN2O3/c1-9(2)16(22)19-11-4-5-14(21)12(8-11)17-20-13-7-10(18)3-6-15(13)23-17/h3-9,20H,1-2H3,(H,19,22)/b17-12-

InChI key

YGMGVHYZWYRTJJ-ATVHPVEESA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Nicotine ditartrate dihydrate EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

related product

Product No.
Description
Pricing

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - STOT RE 1

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alexander Duncan et al.
Nature, 574(7778), 372-377 (2019-10-18)
Diabetes is far more prevalent in smokers than non-smokers, but the underlying mechanisms of vulnerability are unknown. Here we show that the diabetes-associated gene Tcf7l2 is densely expressed in the medial habenula (mHb) region of the rodent brain, where it
Wouter Goutier et al.
Addiction biology, 20(2), 248-258 (2013-11-21)
The present study focused on the evaluation of behavioral sensitization and cross-sensitization induced by nicotine and varenicline in rats. Furthermore, it examined the influence of varenicline, a partial alpha4beta2 nicotinic receptor agonist, on nicotine-induced sensitization. To assess the development of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service