Skip to Content
Merck
All Photos(1)

Key Documents

NIST2383A

Baby food composite

NIST® SRM® 2383a

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24
Product NIST2383A is not currently sold in your country. Contact Technical Service

grade

NMR reference standard

Quality Level

form

solid

packaging

pkg of 4 x 70 g

manufacturer/tradename

NIST®

suitability

suitable for NMR (reference standard)

General description

This Standard Reference Material (SRM) is intended primarily for use in validating methods for determining proximates, calories, vitamins, carotenoids, and elements in food matrices. This SRM can also be used for quality assurance when assigning values to in-house control materials. The baby food composite is a mixture of fruits, vegetables, macaroni, rice flour, and milk powder. For more information, please refer to the SDS and COA.

SRM 2383A_cert

SRM 2383A_SDS

Other Notes

Certified for the analytes listed below. See certificate for values and more details
Ash, Calories, Carbohydrates, Total Sugars, Total Dietary Fiber, Phosphorus (P), Protein, Solids
Carbohydrates: Fructose, Glucose, Lactose, Sucrose
Trace Elements: Arsenic (As), Barium (Ba), Calcium (Ca), Chromium (Cr), Cobalt (Co), Copper (Cu), Iodine (I), Iron (Fe), Magnesium (Mg), Manganese (Mn), Molybdenum (Mo), Nickel (Ni), Potassium (K), Selenium (Se), Sodium (Na), Strontium (Sr), Tin (Sn), Zinc (Zn)
Vitamins: 9-cis-β-Carotene, Free Pantothenic acid (Vitamin B5), Free Vitamin B3 as Niacinamide, Free Vitamin B6 as Pyridoxine, Niacin (Vitamin B3), Niacinamide (Vitamin B3), Pyridoxamine (Vitamin B6), Pyridoxine (Vitamin B6), Riboflavin (Vitamin B2), Thiamine (Vitamin B1), Total Lutein, Total Lycopene, Total Vitamin B5 (Pantothenic acid), Total Vitamin B6 (Pyridoxine hydrochloride), Total Zeaxanthin, Total a-Carotene, Total β-Carotene, Total β-Cryptoxanthin(, trans-Lycopene, trans-β-Carotene

Matrix Group: Processed Food

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service