Skip to Content
Merck
All Photos(1)

Key Documents

NIST1265A

Electrolytic iron

NIST® SRM® 1265a

Sign Into View Organizational & Contract Pricing

Select a Size

1 DISC
$1,410.00

$1,410.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 DISC
$1,410.00

About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

$1,410.00


Check Cart for Availability

Request a Bulk Order

grade

certified reference material

Quality Level

packaging

pkg of disk

manufacturer/tradename

NIST®

format

matrix material

Looking for similar products? Visit Product Comparison Guide

General description

A single unit of SRM 1265A contains a disk measuring 31 mm in diameter and 19 mm in thickness. For more information, please refer to the SDS and COA.
SRM 1265A_cert
SRM 1265A_SDS

Application

Standard Reference Material (SRM) is a high-purity form of electrolytic iron primarily intended for the evaluation of methods for the analysis of elements in steel alloys of similar composition.

Features and Benefits

  • NIST certified values for constituents are provided, which are metrologically traceable to the derived SI unit of mass fraction, expressed in percent (%). · Information values for constituents are listed as mass fractions of the total elements in a steel matrix.
  • Expiration certification, instructions for handling, storage, and use are provided on the NIST certificate.

Other Notes

Certified for the analytes listed below.
Boron (B), Carbon (C), Chromium (Cr), Cobalt (Co), Copper (Cu), Lead (Pb), Manganese (Mn), Molybdenum (Mo), Nickel (Ni), Phosphorus (P), Silicon (Si), Sulfur (S), Vanadium (V)

Reference/Informational value is provided for the following analytes.
Aluminum (Al), Oxygen (O), Antimony (Sb), Silver (Ag), Arsenic (As), Tin (Sn), Germanium (Ge), Titanium (Ti), Hydrogen (H), Tungsten (W), Iron (Fe), Zinc (Zn), Nitrogen (N)

See certificate for values and more details at nist.gov/SRM.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

related product

Product No.
Description
Pricing

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Sol. 2 - Self-heat. 2

Storage Class Code

4.2 - Pyrophoric and self-heating hazardous materials

WGK

nwg

Flash Point(F)

69.8 °F

Flash Point(C)

21 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service