Skip to Content
Merck
All Photos(1)

Documents

NIST2036

Near infrared wavelength/wavenumber reflection standard

NIST® SRM® 2036

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.24

grade

certified reference material

Quality Level

form

solid

packaging

pkg of each

manufacturer/tradename

NIST®

application(s)

general analytical

General description

This Standard Reference Material (SRM) is a certified transfer standard intended for the verification and calibration of wavelength (λ) and wavenumber (μ) scale near-infrared (NIR) spectrometers operating in diffuse reflectance mode. SRM 2036 is a combination of a glass physically contacted with a piece of sinteredpolytetrafluoroethylene (PTFE). A unit of SRM 2036 consists of the optical filter-PTFE assembly mounted in an optical holder, contained in a wooden box. For more information, please refer ot the COA and SDS.

SRM 2036_cert

SRM 2036_SDS

Other Notes

Example analytes are listed below as a reference. Please download a current certificate at nist.gov/SRM for current analytes and certified values.
Holmium oxide , Samarium oxide, Ytterbium oxide, Neodymium oxide, Lanthanum oxide, Boron oxide, Silicon oxide, Zirconium oxide

See certificate for values and more details at nist.gov/SRM.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

related product

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service