Skip to Content
Merck
All Photos(1)

Key Documents

NISTRM8420

Electrolytic iron

NIST® RM 8420

Sign Into View Organizational & Contract Pricing

Select a Size

1 EACH
$844.00

$844.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 EACH
$844.00

About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

$844.00


Check Cart for Availability

Request a Bulk Order

grade

certified reference material

Quality Level

manufacturer/tradename

NIST®

format

matrix material

Other Notes

Please download a current certificate at nist.gov/SRM for current analytes and certified values.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Sol. 2 - Self-heat. 2

Storage Class Code

4.2 - Pyrophoric and self-heating hazardous materials

WGK

nwg

Flash Point(F)

69.8 °F

Flash Point(C)

21 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service