Skip to Content
Merck
All Photos(1)

Documents

86910

Sigma-Aldrich

Titanium(IV) butoxide

purum, ≥97.0% (gravimetric)

Synonym(s):

Orthotitanic acid tetrabutylester, TNBT, TYZOR® TBT organic titanate, Tetrabutyl orthotitanate, Tetrabutyl titanate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Ti(OCH2CH2CH2CH3)4
CAS Number:
Molecular Weight:
340.32
Beilstein:
4148236
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

grade

purum

Quality Level

Assay

≥97.0% (gravimetric)

form

liquid
liquid

reaction suitability

core: titanium
reagent type: catalyst

refractive index

n20/D 1.491 (lit.)

bp

206 °C/10 mmHg (lit.)

density

1.00 g/mL at 20 °C (lit.)

SMILES string

CCCCO[Ti](OCCCC)(OCCCC)OCCCC

InChI

1S/4C4H9O.Ti/c4*1-2-3-4-5;/h4*2-4H2,1H3;/q4*-1;+4

InChI key

YHWCPXVTRSHPNY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Titanium(IV) butoxide is a sourcing material for the preparation of titanium oxide (TiO2) which can further be used in a variety of applications such as dye sensitized solar cells (DSSCs), photo-catalytic and self-cleaning based coatings.
Used to prepare nanosized titania in the anatase form and ferroelectric Bi4Ti3O12 thin films. Used in the preparation of nanocrystalline TiO2 powders at room temperature.

Legal Information

TYZOR is a registered trademark of E. I. du Pont de Nemours and Company

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system, Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

107.6 °F - Pensky-Martens closed cup

Flash Point(C)

42 °C - Pensky-Martens closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Structural and electrical properties of TiO2 thin film derived from sol-gel method using titanium (IV) butoxide
Senain I, et al.
International Journal of Integrated Engineering, 2(3) (2010)
Synthesis and characterization of CNT/TiO 2 composites thermally derived from MWCNT and titanium (IV) n-butoxide
Oh W and Chen M
Bulletin of the Korean Chemical Society,, 29(1), 159-164 (2008)
Preparation and characterization of low temperature sintering nanocrystalline TiO2 prepared via the sol-gel method using titanium (IV) butoxide applicable to flexible dye sensitized solar cells
Bazargan MH, et al.
International Journal of Materials Research, 103(3), 347-351 (2012)
S D Makeev et al.
Klinicheskaia khirurgiia, (1)(1), 19-20 (1989-01-01)
By the method of tissue culture, it is established that the capron threads stained by brilliant green, coated with the chemosorbed substances on the base of titanium alkoxides, polyalkylhydridsiloxanes and polyorganilsiloxanes have no histotoxic properties and can be used in
S Anantharaj et al.
Biomacromolecules, 13(8), 2446-2455 (2012-06-21)
A new dual ester-urethane melt condensation methodology for biological monomers-amino acids was developed to synthesize new classes of thermoplastic polymers under eco-friendly and solvent-free polymerization approach. Naturally abundant L-amino acids were converted into dual functional ester-urethane monomers by tailor-made synthetic

Articles

The prevailing strategies for heat and electric-power production that rely on fossil and fission fuels are having a negative impact on the environment and on our living conditions.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service