Skip to Content
Merck
All Photos(1)

Key Documents

257923

Sigma-Aldrich

Perfluorobutyryl chloride

98%

Synonym(s):

Heptafluorobutyryl chloride

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$61.80
25 G
$180.00

$61.80


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$61.80
25 G
$180.00

About This Item

Linear Formula:
CF3CF2CF2COCl
CAS Number:
Molecular Weight:
232.48
Beilstein:
1790269
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$61.80


Check Cart for Availability

Request a Bulk Order

vapor pressure

7.22 psi ( 20 °C)

Quality Level

Assay

98%

form

liquid

refractive index

n20/D 1.3 (lit.)

bp

39 °C (lit.)

density

1.556 g/mL at 25 °C (lit.)

functional group

acyl chloride
fluoro

storage temp.

2-8°C

SMILES string

FC(F)(F)C(F)(F)C(F)(F)C(Cl)=O

InChI

1S/C4ClF7O/c5-1(13)2(6,7)3(8,9)4(10,11)12

InChI key

WFELVFKXQJYPSL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Perfluorobutyryl chloride has been used in the preparation of:
  • ethylcellulose perfluorobutyrate, non-thrombogenic novel fluoropolymer for gas-exchange membranes[1]
  • 2-perfluoroalkylbenzothiazole[2]

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R J Petersen et al.
Transactions - American Society for Artificial Internal Organs, 21, 242-248 (1975-01-01)
A novel fluoropolymer prepared from ethylcellulose by acylation with perfluorobutyryl chloride has been shown to be remarkably non-thrombogenic in vena cava and renal embolus ring implant tests. Gas permeabilities of this fluoropolymer are dufficient to make it highly attractive for
Synthesis of 2-and 2, 5-substituted benzoxazoles and benzothiazoles.
Soloski EJ, et al.
Journal of Fluorine Chemistry, 8(4), 295-304 (1976)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service