Skip to Content
Merck
All Photos(1)

Key Documents

8.21084

Sigma-Aldrich

Tetra-n-butyl orthotitanate

for synthesis

Synonym(s):

Tetra-n-butyl orthotitanate

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
$45.10
500 ML
$147.00
10 KG
$756.00
40 KG
$2,420.00

$45.10


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
100 ML
$45.10
500 ML
$147.00
10 KG
$756.00
40 KG
$2,420.00

About This Item

Linear Formula:
(CH3CH2CH2CH2O)4Ti
CAS Number:
Molecular Weight:
340.32
MDL number:
UNSPSC Code:
12352103
EC Index Number:
227-006-8
NACRES:
NA.22
grade:
for synthesis
synthesis grade
form:
liquid

$45.10


Estimated to ship on28 May 2025


Request a Bulk Order

grade

for synthesis
synthesis grade

Quality Level

vapor pressure

5.6 hPa ( 20 °C)

form

liquid

potency

3122 mg/kg LD50, oral (Rat)

expl. lim.

2.0-12.0 % (v/v)

bp

310-314 °C/1013 hPa

mp

-40 °C

transition temp

flash point 47 °C

density

0.99 g/cm3 at 20 °C

storage temp.

2-8°C

SMILES string

[Ti](OCCCC)(OCCCC)(OCCCC)OCCCC

InChI

1S/4C4H9O.Ti/c4*1-2-3-4-5;/h4*2-4H2,1H3;/q4*-1;+4

InChI key

YHWCPXVTRSHPNY-UHFFFAOYSA-N

Related Categories

Application

  • Synthesis of TiO2–SiO2: Discusses the use of Tetra-n-butyl orthotitanate in the sol-gel synthesis of TiO2–SiO2, applied as a photocatalyst, emphasizing its utility in creating effective materials for environmental applications (Kien et al., 2023).
  • PbTiO3/PbFe12O19 Nanocomposites: Explores the green synthesis of PbTiO3/PbFe12O19 nanocomposites using Tetra-n-butyl orthotitanate, highlighting an eco-friendly approach to materials synthesis (Ansari, Sobhani, Salavati-Niasari, 2016).

Analysis Note

Assay (ex Ti): ≥ 98.0 %
Density (d 20 °C/ 4 °C): 0.990 - 1.000
Identity (IR): passes test

Signal Word

Danger

Hazard Classifications

Eye Dam. 1 - Flam. Liq. 3 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

116.6 °F

Flash Point(C)

47 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service