Skip to Content
Merck
All Photos(2)

Key Documents

76737

Supelco

3-Ferrocenylpropionic anhydride

for HPLC derivatization, LiChropur, ≥98.0% (C)

Synonym(s):

1,1′′-[Oxybis(3-oxo-3,1-propanediyl)]bis-ferrocene

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$289.00

$289.00


Estimated to ship on26 April 2025


Request a Bulk Order

Select a Size

Change View
100 MG
$289.00

About This Item

Empirical Formula (Hill Notation):
C26H26Fe2O3
CAS Number:
Molecular Weight:
498.17
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:
NACRES:
NA.22

$289.00


Estimated to ship on26 April 2025


Request a Bulk Order

grade

for HPLC derivatization

Quality Level

Assay

≥98.0% (C)

form

crystals

quality

LiChropur

technique(s)

HPLC: suitable

SMILES string

[Fe].[Fe].[CH]1[CH][CH][CH][CH]1.[CH]2[CH][CH][CH][CH]2.O=C(CC[C]3[CH][CH][CH][CH]3)OC(=O)CC[C]4[CH][CH][CH][CH]4

InChI

1S/C16H16O3.2C5H5.2Fe/c17-15(11-9-13-5-1-2-6-13)19-16(18)12-10-14-7-3-4-8-14;2*1-2-4-5-3-1;;/h1-8H,9-12H2;2*1-5H;;

InChI key

HFIVHNMBVHEMJQ-UHFFFAOYSA-N

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

László Szabó et al.
Materials (Basel, Switzerland), 12(1) (2019-01-10)
While intensive efforts are made to prepare carbon fiber reinforced plastics from renewable sources, less emphasis is directed towards elaborating green approaches for carbon fiber surface modification to improve the interfacial adhesion in these composites. In this study, we covalently

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service