Skip to Content
Merck
All Photos(1)

Key Documents

681571

Sigma-Aldrich

(6-Bromohexyl)ferrocene

Synonym(s):

1-Bromo-6-ferrocenylhexane, 6-Ferrocenylhexyl bromide

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
$117.00
1 G
$378.00

$117.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
250 MG
$117.00
1 G
$378.00

About This Item

Empirical Formula (Hill Notation):
C16H21BrFe
CAS Number:
Molecular Weight:
349.09
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

$117.00


Check Cart for Availability

Request a Bulk Order

form

solid

Quality Level

reaction suitability

core: iron
reagent type: catalyst

SMILES string

[Fe].[CH]1[CH][CH][CH][CH]1.BrCCCCCC[C]2[CH][CH][CH][CH]2

InChI

1S/C11H16Br.C5H5.Fe/c12-10-6-2-1-3-7-11-8-4-5-9-11;1-2-4-5-3-1;/h4-5,8-9H,1-3,6-7,10H2;1-5H;

InChI key

LNVDAJXWPKYLFP-UHFFFAOYSA-N

Application

A ferrocene building block.[1][2]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Yongbin Han et al.
Journal of the American Chemical Society, 128(42), 13913-13920 (2006-10-19)
This work reports a new biocompatible surfactant structure, of which the hydrophilic head group is composed of a folded, stable self-inclusion complex of a ferrocenyl substituted beta-cyclodextrin (betaCD). While multiple intra- or intermolecular complexes can exist for this amphiphile, the
Eric Bakker
Journal of electroanalytical chemistry (Lausanne, Switzerland), 639(1-2), 1-7 (2010-08-10)
A generalized description of the response behavior of potentiometric polymer membrane ion-selective electrodes is presented on the basis of ion-exchange equilibrium considerations at the sample-membrane interface. This paper includes and extends on previously reported theoretical advances in a more compact

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service