Skip to Content
Merck
All Photos(3)

Key Documents

901699

Sigma-Aldrich

3-Mercaptopropanyl-N-hydroxysuccinimide ester

>95%

Synonym(s):

NHS ester

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$127.00
5 G
$539.00

$127.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$127.00
5 G
$539.00

About This Item

Empirical Formula (Hill Notation):
C7H9NO4S
CAS Number:
Molecular Weight:
203.22
MDL number:
UNSPSC Code:
12352005
NACRES:
NA.23

$127.00


Check Cart for Availability

Request a Bulk Order

Assay

>95%

form

solid or liquid

reaction suitability

reagent type: linker

color

colorless to yellow

functional group

NHS ester
thiol

storage temp.

2-8°C

SMILES string

SCCC(ON1C(CCC1=O)=O)=O

InChI

1S/C7H9NO4S/c9-5-1-2-6(10)8(5)12-7(11)3-4-13/h13H,1-4H2

InChI key

UTBOEPMMEANMPS-UHFFFAOYSA-N

Application

3-Mercaptopropanyl-N-hydroxysuccinimide ester is an aliphatic, heterobifunctional crosslinking reagent. Due to the functional groups, this material can be used in a multitude of applications, such as the synthesis of protien-polymer conjugates and the synthesis of functionalized quantum dots. In addition to use as a crosslinking agent, this material can also be used to install functional groups post-polymerization. The terminal N-hydroxysuccinimide (NHS) ester can react with amines while the terminal thiol can react with thiols, alkenes, and alkynes. The free thiol can also be used to form self-assembled monolayers (SAMs) on gold surfaces.
3-Mercaptopropanyl-N-hydroxysuccinimide ester can be used in the synthesis of diaminobutane (DAB) based dendrimers for the formation of an ultrafiltration membrane for the removal of heavy metals.[1][2]

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

110.0 °F

Flash Point(C)

43.33 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Inclusion of thiol DAB dendrimer/CdSe quantum dots based in a membrane structure: Surface and bulk membrane modification
Algarra M, et al.
Electrochimica Acta, 89, 652-659 (2013)
Characterization of an engineered cellulose based membrane by thiol dendrimer for heavy metals removal
Algarra M, et al.
Chemical Engineering Journal, 253, 472-477 (2014)

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service