Skip to Content
Merck
All Photos(1)

Key Documents

74595

Supelco

(±)-Glycidol

analytical standard

Synonym(s):

(±)-Oxirane-2-methanol, 2,3-Epoxy-1-propanol, Glycerolglycide

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$86.50

$86.50


Estimated to ship on01 June 2025


Request a Bulk Order

Select a Size

Change View
100 MG
$86.50

About This Item

Empirical Formula (Hill Notation):
C3H6O2
CAS Number:
Molecular Weight:
74.08
Beilstein:
383562
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

$86.50


Estimated to ship on01 June 2025


Request a Bulk Order

grade

analytical standard

Quality Level

vapor density

2.15 (vs air)

vapor pressure

0.9 mmHg ( 25 °C)

Assay

≥95.0% (GC)

autoignition temp.

780 °F

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.429-1.437
n20/D 1.433 (lit.)

bp

61-62 °C/15 mmHg (lit.)

density

1.117 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

SMILES string

OCC1CO1

InChI

1S/C3H6O2/c4-1-3-2-5-3/h3-4H,1-2H2

InChI key

CTKINSOISVBQLD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Glycidol is an aliphatic epoxide[1] which may be used as a stabilizer in the manufacture of vinyl polymers, as an additive for oil and synthetic hydraulic fluids, and also as a diluent in some epoxy resins.[2]

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Carc. 1B - Eye Irrit. 2 - Muta. 2 - Repr. 1B - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

161.6 °F

Flash Point(C)

72 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Carcinogenicity of glycidol in F344 rats and B6C3F1 mice.
Irwin RD, et al.
Journal of Applied Toxicology, 16(3), 201-209 (1996)
NTP Toxicology and Carcinogenesis Studies of Glycidol (CAS No. 556-52-5) In F344/N Rats and B6C3F1 Mice (Gavage Studies).
National Toxicology Program and others
National Toxicology Program Technical Report Series, 374(3), 1-1 (1990)
Hirotoshi Akane et al.
Journal of applied toxicology : JAT, 34(12), 1389-1399 (2014-01-08)
We previously found that exposure to glycidol at 1000 ppm in drinking water caused axonopathy in maternal rats and aberrations in late-stage hippocampal neurogenesis, targeting the process of neurite extension in offspring. To identify the profile of developmental neurotoxicity of
Glycidol.
IARC monographs on the evaluation of carcinogenic risks to humans, 77, 469-486 (2000-12-02)
Nadiya Bakhiya et al.
Molecular nutrition & food research, 55(4), 509-521 (2011-02-26)
Fatty acid esters of 3-chloropropane-1,2-diol (3-MCPD) and glycidol are a newly identified class of food process contaminants. They are widespread in refined vegetable oils and fats and have been detected in vegetable fat-containing products, including infant formulas. There are no

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service