Skip to Content
Merck
All Photos(1)

Key Documents

614661

Sigma-Aldrich

3-Chloro-1,2-propanediol-1,1,2,3,3-d5

98 atom % D, 97% (CP)

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$1,490.00

$1,490.00


Check Cart for Availability

Request more information

Select a Size

Change View
100 MG
$1,490.00

About This Item

Linear Formula:
ClCD2CD(OH)CD2OH
Molecular Weight:
115.57
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:

$1,490.00


Check Cart for Availability

Request more information

isotopic purity

98 atom % D

Quality Level

Assay

97% (CP)

bp

216-219 °C

density

1.381 g/mL at 25 °C

mass shift

M+5

SMILES string

[2H]C([2H])(O)C([2H])(O)C([2H])([2H])Cl

InChI

1S/C3H7ClO2/c4-1-3(6)2-5/h3,5-6H,1-2H2/i1D2,2D2,3D

InChI key

SSZWWUDQMAHNAQ-UXXIZXEISA-N

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Oral - Carc. 2 - Eye Dam. 1 - Muta. 2 - Repr. 1B

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thorsten Buhrke et al.
Archives of toxicology, 89(12), 2243-2251 (2014-10-31)
The food contaminants 3-chloro-1,2-propanediol (3-MCPD) and 3-MCPD fatty acid esters have attracted considerable attention in the past few years due to their toxic properties and their occurrence in numerous foods. Recently, significant amounts of the isomeric compounds 2-chloro-1,3-propanediol (2-MCPD) fatty
Roberta Andreoli et al.
Analytical and bioanalytical chemistry, 407(16), 4823-4827 (2015-04-12)
A new method for the simultaneous quantitative determination in human urine of 3-monochloropropane-1,2-diol (3-MCPD), a toxic food contaminant, and its metabolite, 2,3-dihydroxypropyl mercapturic acid (DHPMA), was developed and validated. After urine dilution, the analytes were separated on an Atlantis®dC18 column

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service