Skip to Content
Merck
All Photos(1)

Documents

16761

Supelco

2-Chloro-1,3-propanediol

analytical standard

Synonym(s):

2-MCPD, 2-Monochloro-propanediol, Glycerol β-chlorohydrin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H7ClO2
CAS Number:
Molecular Weight:
110.54
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥98.0% (GC)

shelf life

limited shelf life, expiry date on the label

storage temp.

2-8°C

SMILES string

OCC(Cl)CO

InChI

1S/C3H7ClO2/c4-3(1-5)2-6/h3,5-6H,1-2H2

InChI key

DYPJJAAKPQKWTM-UHFFFAOYSA-N

Related Categories

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service