Skip to Content
Merck
All Photos(3)

Key Documents

71007ATMBC

Millipore

Assurance® GDS E. coli O157:H7 Tq Kit for Automation

suitable for PCR testing in Food and Beverages applications

Synonym(s):

Nucleic acid amplification system

Sign Into View Organizational & Contract Pricing

Select a Size

576 TESTS
$10,550.00

$10,550.00


Check Cart for Availability

Request more information

Select a Size

Change View
576 TESTS
$10,550.00

About This Item

UNSPSC Code:
41106302
NACRES:
NA.85

$10,550.00


Check Cart for Availability

Request more information

Product Name

GDS E. coli O157:H7 Tq Automation Kit, BioControl, Molecular based PCR test for the detection of E. coli O157:H7 in food and environmental samples

Agency

PCR, Immunomagnetic Separation (IMS)
AOAC® (OMA)
AOAC® (PTM)
Health Canada
MicroVal

Quality Level

product line

Assurance GDS®

shelf life

6‑12 mo.

packaging

pkg of 576

manufacturer/tradename

BioControl Systems 71007-576ATM

technique(s)

PCR: suitable

application(s)

food and beverages
pathogen testing

compatibility

BioControl GDS
for use with GDS Rotor-Gene

storage temp.

2-8°C

suitability

Escherichia coli (O157:H7)

General description

GDS E. coli O157:H7 Tq Automation Kit is an automated nucleic acid amplification system for the detection of E. coli O157:H7 in food and environmental samples. The proprietary PickPen® IMS-based sample preparation procedure quickly and effectively captures E. coli O157:H7, separating the pathogen from inhibitory background materials and ensuring valid PCR reactions for all sample types, including challenging food and environmental samples. Assurance® GDS PCR technology provides two additional levels of specificity using both highly specific primers and probes to ensure accurate detection of E. coli O157:H7.

Application

GDS for E. coli O157:H7 Tq is an automated nucleic acid amplification system for the detection of pathogenic E. coli O157:H7 in foods, ingredients, and environmental samples.

Features and Benefits

  • Results available as fast as 8 hours for 25 g samples / 10 hours for composite 375 g samples
  • Positive samples can be confirmed for stx genes in an additional 2 hours with GDS Shiga Toxin Genes (O157) assay (PN 71005BC).
  • Validated for raw ground beef, raw beef trim, frozen finely textured beef, orange juice, apple juice, fresh vegetables, sprout process water, carcass cloths, among others
  • Features innovative PickPen® immunomagnetic separation (IMS) for additional specificity and fast time to results
  • Simplified sample prep does not require heating blocks or preparation of reagents

Components

Each 576ATM (for PPMX Automation) kit includes:
  • E. coli O157:H7 Amplification Tubes Tq (0.2 mL)
  • O157 Concentration Reagent

For each 576ATM kit, the following are also necessary and should be ordered separately:
  • 61031BC Wash Solution Kit
  • 34724BC Resuspension Buffer Tq

Testing Method

PCR, Immunomagnetic Separation (IMS)

Legal Information

AOAC is a registered trademark of AOAC International
Assurance is a registered trademark of Merck KGaA, Darmstadt, Germany
Assurance GDS is a registered trademark of BioControl Systems, Inc.
PICKPEN is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service