Skip to Content
Merck
All Photos(1)

Key Documents

Z273554

Sigma-Aldrich

Molykote high-vacuum silicone grease

colorless, weight 5.3 oz (tube)

Synonym(s):

High-Vacuum Silicone Grease, Molykote Grease, Vacuum Grease

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$111.00

$111.00


Available to ship on28 April 2025Details



Select a Size

Change View
1 EA
$111.00

About This Item

UNSPSC Code:
15121900
NACRES:
NB.35

$111.00


Available to ship on28 April 2025Details


material

colorless

weight

5.3 oz (tube)

General description

Seals and prevents freezing of stopcocks and ground-glass joints in high-vacuum systems at pressures less than 10-6 mm Hg. Heat stable (−40 to 204 °C), low vapor pressure, and chemically resistant. Colorless. 5.3 oz. tube.

Application

Dow Corning® high-vacuum silicone grease has been used:
  • to coat the cloning rings used for hypoxanthine phosphoribosyltransferase (HPRT)-deficient cells selection and isolation
  • to coat the cloning rings used for selecting the transformed cells during BALB/c cell transformation assay
  • to seal a cover glass over a petri dish containing culture for bioluminescence detection

Legal Information

Corning is a registered trademark of Corning, Inc.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yulia Komarova et al.
The Journal of cell biology, 184(5), 691-706 (2009-03-04)
End binding proteins (EBs) are highly conserved core components of microtubule plus-end tracking protein networks. Here we investigated the roles of the three mammalian EBs in controlling microtubule dynamics and analyzed the domains involved. Protein depletion and rescue experiments showed
Mitra Aliabouzar et al.
Soft matter, 16(28), 6501-6513 (2020-07-01)
Acoustically-responsive scaffolds (ARSs), which are composite fibrin hydrogels, have been used to deliver regenerative molecules. ARSs respond to ultrasound in an on-demand, spatiotemporally-controlled manner via a mechanism termed acoustic droplet vaporization (ADV). Here, we study the ADV-induced, time-dependent micromechanical and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service