Skip to Content
Merck
All Photos(1)

Key Documents

1.18755

Supelco

Ozone Test Kit, colorimetric

0.007-0.20 mg/L (O₃), MQuant®

Synonym(s):

Ozone Visual Test Kit

Sign Into View Organizational & Contract Pricing

Select a Size

300 TESTS
$289.00

$289.00


Estimated to ship on31 May 2025


Request a Bulk OrderRequest more information

Select a Size

Change View
300 TESTS
$289.00

About This Item

UNSPSC Code:
41116116
NACRES:
NA.24

$289.00


Estimated to ship on31 May 2025


Request a Bulk OrderRequest more information

Product Name

Ozone Test, colorimetric, 0.007-0.20 mg/L (O3), for use with MCOLORTEST®

product line

MColortest®

Quality Level

specific analyte(s)

ozone

measuring range

0.007-0.20 mg/L (O3)

compatibility

for use with MCOLORTEST®

detection method

colorimetric

storage temp.

15-25°C

General description

Method: colorimetric, DPD, with color card and comparator block with long test tubes 0.007 - 0.017 - 0.030 - 0.040 - 0.055 - 0.070 - 0.10 - 0.14 - 0.20 mg/l O2 MQuant®
The MQuant® Ozone test (formally MColortest®) measures the content of ozone in:
  • Swimming-pool water
  • Drinking water
  • Disinfectant solutions
  • Process water
  • Wastewater
This test is not suited for seawater.

Method
In weakly acidic solution ozone reacts with dipropyl-p-phenylenediamine (DPD) to form a red-violet dye. The ozone concentration is measured semiquantitatively by visual comparison of the color of the measurement solution with the color fields of a color card. The unique brilliance of the print and their fine color graduation allow precise analyses, even at the lower end of the concentration range. Due to the long test tubes a sensitive measurement range can be covered.
Using the refill pack 1.18759 containing all the reagents, the original test kit can be used for a very long time, making it a particularly economical and ecological system. 

Legal Information

MCOLORTEST is a registered trademark of Merck KGaA, Darmstadt, Germany
MQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation markHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Repr. 1B - STOT RE 2 Oral

Target Organs

Thyroid

Storage Class Code

6.1D - Non-combustible, acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service