Skip to Content
Merck
All Photos(2)

Key Documents

66761

Supelco

Ethyl cinnamate

analytical standard

Synonym(s):

3-Phenyl-2-propenoic acid ethyl ester, Ethyl 3-phenyl-2-propenoate, NSC 6773

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH=CHCOOC2H5
CAS Number:
Molecular Weight:
176.21
Beilstein:
775541
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥98.5% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

≤0.3% water

refractive index

n20/D 1.558 (lit.)
n20/D 1.559-1.561

bp

271 °C (lit.)

mp

6-8 °C (lit.)

density

1.049 g/mL at 20 °C (lit.)

application(s)

cleaning products
cosmetics
environmental
flavors and fragrances
food and beverages
personal care

format

neat

storage temp.

2-8°C

SMILES string

CCOC(=O)\C=C\c1ccccc1

InChI

1S/C11H12O2/c1-2-13-11(12)9-8-10-6-4-3-5-7-10/h3-9H,2H2,1H3/b9-8+

InChI key

KBEBGUQPQBELIU-CMDGGOBGSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Ethyl cinnamate may be used as a reference standard:
  • to determine the toxic effects of ethyl cinnamate, affecting the growth, photosynthesis and cell physiology of Chlorella vulgaris, a dominant species of green algae based on chlorophyll fluorescence and flow cytometry analysis.
  • in the determination of ethyl cinnamate in samples of red wine using gas chromatography coupled with ion-trap mass spectrometric analysis technique (GC-MS).
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Comparative study of batch and fluidized bed bioreactors for lipase-catalyzed ethyl cinnamate synthesis.
Industrial & Engineering Chemistry Research, 52(47), 16689-16697 (2013)
Bingham E and Cohrssen B
Patty's Toxicology (2012)
Toxic effects of ethyl cinnamate on the photosynthesis and physiological characteristics of Chlorella vulgaris based on chlorophyll fluorescence and flow cytometry analysis.
Jiao Y, et al.
TheScientificWorldJournal (2015)
Quantitative determination of trace and ultratrace flavour active compounds in red wines through gas chromatographic?ion trap mass spectrometric analysis of microextracts.
Ferreira V, et al.
Journal of Chromatography A, 806(2), 349-354 (1998)
S P Bhatia et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 45 Suppl 1, S90-S94 (2007-11-27)
A toxicologic and dermatologic review of ethyl cinnamate when used as a fragrance ingredient is presented.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service