Skip to Content
Merck
All Photos(1)

Key Documents

W346209

Sigma-Aldrich

Maltyl isobutyrate

≥98%, FG

Synonym(s):

Maltol isobutyrate

Sign Into View Organizational & Contract Pricing

Select a Size

W346209-SAMPLE-K
$94.30
100 G
$126.00
1 KG
$421.00

$94.30


Estimated to ship on15 April 2025


Request a Bulk OrderRequest more information

Select a Size

Change View
W346209-SAMPLE-K
$94.30
100 G
$126.00
1 KG
$421.00

About This Item

Empirical Formula (Hill Notation):
C10H12O4
CAS Number:
Molecular Weight:
196.20
FEMA Number:
3462
EC Number:
Council of Europe no.:
10739
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.525
NACRES:
NA.21
Organoleptic:
caramel; jam; fruity; sweet
grade:
FG
Halal
Kosher
biological source:
synthetic
Agency:
meets requirements for JECFA
food allergen:
no known allergens

$94.30


Estimated to ship on15 April 2025


Request a Bulk OrderRequest more information

biological source

synthetic

Quality Level

grade

FG
Halal
Kosher

Agency

meets requirements for JECFA

reg. compliance

EU Regulation 1334/2008 & 178/2002

Assay

≥98%

refractive index

n20/D 1.497 (lit.)

density

1.149 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

caramel; jam; fruity; sweet

SMILES string

CC(C)C(=O)OC1=C(C)OC=CC1=O

InChI

1S/C10H12O4/c1-6(2)10(12)14-9-7(3)13-5-4-8(9)11/h4-6H,1-3H3

InChI key

VSBHYRPUJHEOBE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Maltyl isobutyrate is a flavoring agent used in the food industry.[1] It is one of the constituents of cigarette smoke.[2]

Biochem/physiol Actions

Taste at 10-30 ppm

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dictionary of Food Compounds with CD-ROM: Additives, Flavors, and Ingredients, 1013-1013 (2012)
An overview of the effects of tobacco ingredients on smoke chemistry and toxicity.
Baker RR, et al.
Food And Chemical Toxicology, 42, 53-83 (2004)
The effect of tobacco ingredients on smoke chemistry. Part I: Flavourings and additives.
Baker RR, et al.
Food And Chemical Toxicology, 42, 3-37 (2004)
Mosciano, G.
Perfumer & Flavorist, 25, 72-72 (2000)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service