Skip to Content
Merck
All Photos(1)

Key Documents

F2793

Supelco

D-(−)-Fructose

analytical standard, analytical standard for fructose assay kit, for use with enzymatic assay kit FA20

Synonym(s):

D-Levulose, Fruit sugar

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
$30.00

$30.00


Available to ship on09 April 2025Details


Request a Bulk Order

Select a Size

Change View
500 MG
$30.00

About This Item

Empirical Formula (Hill Notation):
C6H12O6
CAS Number:
Molecular Weight:
180.16
Beilstein:
1239004
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

$30.00


Available to ship on09 April 2025Details


Request a Bulk Order

biological source

corn

Quality Level

grade

analytical standard

Assay

≥99% (HPLC)

analyte chemical class(es)

oligosaccharides

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

119-122 °C (dec.) (lit.)

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

SMILES string

OC[C@@H](O)[C@@H](O)[C@H](O)C(=O)CO

InChI

1S/C6H12O6/c7-1-3(9)5(11)6(12)4(10)2-8/h3,5-9,11-12H,1-2H2/t3-,5-,6-/m1/s1

InChI key

BJHIKXHVCXFQLS-UYFOZJQFSA-N

Looking for similar products? Visit Product Comparison Guide

General description

D-(−)-Fructose is a important constituent of sucrose, found in many plants storage organs, which contributes to fruit sweetness and is a major source of calories in fruit consumption.[1]

Application

D-(−)-Fructose is used as an analytical reference standard for the quantification of the analyte in plant tissues using high-performance liquid chromatography coupled to mass spectrometry.[1]
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Validation of a rapid and sensitive HPLC/MS method for measuring sucrose, fructose and glucose in plant tissues
Georgelis N, et al.
Food Chemistry, 262, 191-198 (2018)
Diamante Maresca et al.
Microorganisms, 7(7) (2019-07-18)
Phenotypic and genotypic evidence indicates that many LAB strains can grow in presence of oxygen and can shift from fermentative to aerobic and/or respiratory metabolism. The aerobic and respiratory growth of several LAB species have been studied, allowing the selection
Kyung-Hyun Cho
Molecules (Basel, Switzerland), 26(14) (2021-07-25)
Beta (β)-amyloid (Aβ) is a causative protein of Alzheimer's disease (AD). In the pathogenesis of AD, the apolipoprotein (apo) A-I and high-density lipoprotein (HDL) metabolism is essential for the clearance of Aβ. In this study, recombinant Aβ42 was expressed and
Carlos A Roncal Jimenez et al.
Kidney international, 86(2), 294-302 (2013-12-18)
The epidemic of chronic kidney disease in Nicaragua (Mesoamerican nephropathy) has been linked with recurrent dehydration. Here we tested whether recurrent dehydration may cause renal injury by activation of the polyol pathway, resulting in the generation of endogenous fructose in
Jérémy Lothier et al.
Planta, 240(3), 629-643 (2014-07-16)
The first 6-fructan exohydrolase (6-FEH) cDNA from Lolium perenne was cloned and characterized. Following defoliation, Lp6 - FEHa transcript level unexpectedly decreased together with an increase in total FEH activity. Lolium perenne is a major forage grass species that accumulates

Questions

1–2 of 2 Questions  
  1. Can we use this fructose standard for bee pollen sugar(fructose) hplc analysis? OR there is another analytical fructose standard for bee pollen sugars.

    1 answer
    1. Yes, this product is suitable for use in HPLC analysis. However, it is for identity testing and not specific for bee pollen sugar. For content/assay (quantitative) testing, product PHR1002 will be recommended. Please see the link below to review this product option:
      https://www.sigmaaldrich.com/product/sial/phr1002

      Helpful?

  2. Necesitamos meter fructosa en un animalario para incorporarla a la bebida de los animales. ¿Se puede autoclavar fructosa en polvo para esterilizarla?

    1 answer
    1. Yes, this material is suitable for autoclaving. Note that this product is intended for reasearch use only. It is not suitable for human or veterinary use.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service